Learn More
DGCR14 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA551542
Additional Details : Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: ASASSLLLPA ASRPPRKREA GEAGAATSKQ RVLDEEEYIE GLQTVIQRDF FPDVEKLQAQ KEYLEAEENG DLERMRQIAI KFGSALGKMS REPPPPYVTP ATFETPEVHA GTGVVGNKPR PRGRGLEDGE AGEEEEKEPL PSLDV Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 86%.
May be involved in pre-mRNA splicing.Spezifikation
DGCR14 | |
Polyclonal | |
Unconjugated | |
ESS2 | |
AI462402; D16H22S1269E; Dgcr1; DGCR13; DGCR14; DGSH; DGS-H; DGSI; DGS-I; DiGeorge syndrome critical region 13; DiGeorge syndrome critical region 14; diGeorge syndrome critical region 14 homolog; DiGeorge syndrome critical region gene 13; DiGeorge syndrome critical region gene 14; DiGeorge syndrome critical region gene 14 homolog; DiGeorge syndrome critical region gene DGSI; DiGeorge syndrome critical region protein 14; DiGeorge syndrome gene H; DiGeorge syndrome gene I; diGeorge syndrome protein H; ES2; ES2 protein; Es2el; ESS2; Expressed sequence 2 embryonic lethal; Protein DGCR13; protein DGCR14; Protein ES2; Splicing factor ESS-2 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8220 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96DF8 | |
ESS2 | |
Recombinant protein corresponding to Human DGCR14 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only