missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmoglein 3 Monoclonal Antibody (5G11), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen 326300
Additional Details : Gewicht : 0.50000kg
Beschreibung
Desmoglein 3 Monoclonal antibody specifically detects Desmoglein 3 in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Western Blot.Spezifikation
Desmoglein 3 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
P32926 | |
Dsg3 | |
Human desmoglein-3 extracellular domain. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
5G11 | |
Unconjugated | |
Dsg3 | |
130 kDa pemphigus vulgaris antigen; 130 kDa pemphigus vulgaris antigen homolog; bal; balding; Cadherin family member 6; CDHF6; desmoglein 3; desmoglein-3; DKFZp686P23184; Dsg3; PVA | |
Mouse | |
Protein A | |
RUO | |
1830 | |
-20°C | |
Liquid |