Learn More
Abnova™ DEPDC1B Recombinant Protein
Marke: Abnova™ H00055789-P01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Spezifikation
AAH10904 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
46.31 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
MPPLCDGFGTRTLMVQTFSRCILCSKDEVDLDELLAARLVTFLMDNYQEILKVPLALQTSIEERVAHLRRVQVKYPGADMDITLSAPSFCRQISPEEFEYQRSYGSQEPLAALLEEVITDAKLSNKEKKKKLKQFQKSYPEVYQERFPTPESAALLFPEKPKPKPQLLMWALKKPFQPFQRTRSFRM | |
BRCC3/FLJ11252/XTP1 | |
DEPDC1B | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
55789 | |
DEPDC1B (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DEPDC1B | |
Human | |
Recombinant | |
Solution |