Learn More
DENND5A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563049
Additional Details : Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: SALWSHLLHY QDNRQRKLTS GSLSTSGILL DSERRKSDAS SLMPPLRISL IQD Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 91%.
DENND5A Polyclonal antibody specifically detects DENND5A in Human samples. It is validated for Immunohistochemistry.Spezifikation
DENND5A | |
Polyclonal | |
Unconjugated | |
DENND5A | |
1500012B19Rik; AJ245569; AW496491; DENN domain containing 5A; DENN domain-containing protein 5A; DENN/MADD domain containing 5A; DENND5A; KIAA1091; mKIAA1091; ORF37; Rab6 interacting protein 1; rab6-interacting protein 1; RAB6IP1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23258 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6IQ26 | |
DENND5A | |
Recombinant protein corresponding to Human DENND5A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only