missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DENND5A Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA562705
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Immunogen sequence: MHNAEYDVLH APPADDRDQS SMEDGEDTPV TKLQR Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 80%.
DENND5A (DENN domain-containing protein 5A) is a guanine nucleotide exchange factor (GEF) which activates RAB6A and RAB39A/RAB39B. It promotes the exchange of GDP to GTP by converting inactive GDP-bound Rab proteins to their active GTP form. DENND5A is also involved in the negative regulation of neurtie outgrowth. Mutations affecting this gene can result in epileptic encephalopathy, early infantile, 49 (EIEE49).
Spezifikation
| DENND5A | |
| Polyclonal | |
| Unconjugated | |
| DENND5A | |
| 1500012B19Rik; AJ245569; AW496491; DENN domain containing 5A; DENN domain-containing protein 5A; DENN/MADD domain containing 5A; DENND5A; KIAA1091; mKIAA1091; ORF37; Rab6 interacting protein 1; rab6-interacting protein 1; RAB6IP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23258 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6IQ26 | |
| DENND5A | |
| Recombinant protein corresponding to Human DENND5A. Recombinant protein control fragment (Product #RP-100833). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur