Learn More
DCTN5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA559816
Additional Details : Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LGELLYNKSE YIETASGNKV SRQSVLCGSQ NIVLNGKTIV MNDCIIRGDL ANVRVGRHCV VKSRSVIRPP FKKFSKGV Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
This gene encodes a subunit of dynactin, a component of the cytoplasmic dynein motor machinery involved in minus-end-directed transport. The encoded protein is a component of the pointed-end subcomplex and is thought to bind membranous cargo. A pseudogene of this gene is located on the long arm of chromosome 1. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.Spezifikation
DCTN5 | |
Polyclonal | |
Unconjugated | |
DCTN5 | |
4930427E12Rik; b2b315Clo; C78178; dctn5; dynactin 4; dynactin 5; dynactin 5 (p25); dynactin subunit 5; dynactin subunit 5-like protein; dynactin subunit p25; MGC3248 protein; p25; p25 dynactin subunit; RGD1305654; zgc:92842 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84516 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BTE1 | |
DCTN5 | |
Recombinant protein corresponding to Human DCTN5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only