Learn More
DCLRE1A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA557868
Additional Details : Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: TTPGKLCRTQ KSQHVSPKIR PVYDGYCPNC QMPFSSLIGQ TPRWHVFECL DSPPRSETEC PDGLLCTSTI PFHYKRYTHF LLAQSRAG Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 74%.
May be required for DNA interstrand cross-link repair. Also required for checkpoint mediated cell cycle arrest in early prophase in response to mitotic spindle poisons.Spezifikation
DCLRE1A | |
Polyclonal | |
Unconjugated | |
DCLRE1A | |
DCLRE1A; DNA cross-link repair 1A; DNA cross-link repair 1A protein; DNA-crosslink repair gene SNM1; hSNM1; hSNM1A; KIAA0086; PSO2; SNM1; SNM1 homolog A; SNM1A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9937 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.08 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6PJP8 | |
DCLRE1A | |
Recombinant protein corresponding to Human DCLRE1A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only