missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DAPK1, GST Tag Recombinant Protein
DAPK1 Recombinant Human Protein
824.00 CHF
Spezifikation
Zugriffsnummer | NP_004929 |
---|---|
Zur Verwendung mit (Anwendung) | Kinase Assay |
Molekulargewicht | 71.6 kDa |
Gen-Alias | DAPK1 |
Kinase-Familie | DAPK Kinase Family |
Beschreibung
DAPK1 is a large, calmodulin-dependent kinase involved in gamma-interferon induced programmed cell death and is a potential tumour suppressor.Spezifikation
NP_004929 | |
71.6 kDa | |
DAPK Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors |
Kinase Assay | |
DAPK1 | |
Human | |
GST Tag | |
Ser⁄Thr Kinases: CAMK Group | |
Baculovirus |