missing translation for 'onlineSavingsMsg'
Learn More

D2HGDH Mouse anti-Human, Mouse, Clone: 1E9B3, Proteintech

Mouse Monoclonal Antibody

Marke:  Proteintech 66364-1-IG-20UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16838175

  • 133.80 CHF / 20 Mikroliter
Verfügbar ab: 21-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

This gene encodes D-2hydroxyglutarate dehydrogenase, a mitochondrial enzyme belonging to the FAD-binding oxidoreductase/transferase type 4 family. This enzyme, which is most active in liver and kidney but also active in heart and brain, converts D-2-hydroxyglutarate to 2-ketoglutarate. Mutations in this gene are present in D-2-hydroxyglutaric aciduria, a rare recessive neurometabolic disorder causing developmental delay, epilepsy, hypotonia, and dysmorphic features.
Spezifikation

Spezifikation

D2HGDH
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8CIM3, Q8N465
D2HGDH
D2HGDH Fusion Protein Ag5036
20 μL
Primary
Human, Mouse
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
1E9B3
Unconjugated
D2HGDH
D2HGD, D2HGDH
Mouse
Protein G
RUO
728294, 98314
-20°C
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote