Learn More
Melford D-Luciferin Potassium Salt
Artikelnummer. p-200086985
Click to view available options
Menge:
50 mg
100 mg
250 mg
1 g
Packungsgröße:
1 Gramm
100 Milligramm
250 Milligramm
50 Milligramm
Artikelnummer. 30018441
Marke: Melford L370600.25
om dit product te kopen Registreer vandaag om een webaccount aan te maken
RPI offers the highest purity D-Luciferin Potassium Salt for a wide range of bioluminescent applications.
RPI offers the highest purity D-Luciferin Potassium Salt for a wide range of bioluminescent applications. Produces bioluminescence when oxidized by firefly luciferase. Allows highly sensitive bioluminescent measurements. Common research uses reporter gene assays, ATP assays, whole animal studies, DNA sequencing and others. Our product is specially purified and tested extensively to ensure the highest signal and reproducibility with low background and sensitivity. Guaranteed purity is determined by both standard HPLC and Chiral HPLC methods. Protect from light and moisture.
Spezifikation
Menge 250 mg