missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 7 Rabbit, Clone: KRT7/1499R, enQuireBio™
Rabbit Monoclonal Antibody
Marke: EnquireBio 3855-RBM9-P0
Additional Details : Gewicht : 0.01000kg
Spezifikation
Cytokeratin 7 | |
Cytoplasmic | |
KRT7/1499R | |
Unconjugated | |
NP_011536627 | |
Rabbit | |
55kDa | |
Protein A/G Purified | |
RUO | |
3855 | |
IgG κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
KRT7 | |
Recombinant full-length human KRT7 protein | |
HeLa cells. Carcinoma of ovary, lung, cervix, or breast | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |