missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytochrome B5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA5115072
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Antibody detects endogenous levels of total CYB5.
Cytochrome b5 is a membrane-bound member of the cytochrome b family. A heme protein that functions as an electron carrier for many membrane-bound oxygenases, cytochrome b5 possesses two heme groups, which are not covalently attached to the protein. Two isoforms of cytochrome b5, a microsomal membrane-bound form and a cytoplasmic form, are produced by alternative splicing. Mutations in cytochrome b5 are associated with Leber's hereditary optic neuropathy and with myopathy.
Spezifikation
| Cytochrome B5 | |
| Polyclonal | |
| Unconjugated | |
| CYB5A | |
| 0610009N12Rik; Cyb5; CYB5A; cytochrome b5; cytochrome b-5; cytochrome b5 type A; cytochrome b5 type A (microsomal); MCB5; Microsomal cytochrome b5 type A; type 1 cyt-b5 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 109672, 1528, 64001 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P00167, P00173, P56395 | |
| CYB5A | |
| A synthesized peptide derived from human CYB5A(Accession P00167), corresponding to amino acid residues N62-N112. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur