Learn More
Invitrogen™ Cytochrome B5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA527570
Beschreibung
Recommended positive controls: Cytochrome b5 tansfected 293T cell. Predicted reactivity: Mouse (90%), Rat (90%), Xenopus laevis (81%), Dog (96%), Pig (93%), Chicken (82%), Sheep (93%), Rhesus Monkey (97%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Cytochrome b5 is a membrane-bound member of the cytochrome b family. A heme protein that functions as an electron carrier for many membrane-bound oxygenases, cytochrome b5 possesses two heme groups, which are not covalently attached to the protein. Two isoforms of cytochrome b5, a microsomal membrane-bound form and a cytoplasmic form, are produced by alternative splicing. Mutations in cytochrome b5 are associated with Leber's hereditary optic neuropathy and with myopathy.
Spezifikation
| Cytochrome B5 | |
| Polyclonal | |
| Unconjugated | |
| CYB5A | |
| 0610009N12Rik; Cyb5; CYB5A; cytochrome b5; cytochrome b-5; cytochrome b5 type A; cytochrome b5 type A (microsomal); MCB5; Microsomal cytochrome b5 type A; type 1 cyt-b5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1528 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.28 mg/mL | |
| PBS with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P00167 | |
| CYB5A | |
| Recombinant protein encompassing a sequence within the center region of human Cytochrome b5. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.