missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP4F3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 19701-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Cytochrome P450 super family proteins are monooxygenases which catalyze multiple biochemical reactions involved in drug metabolism and synthesis of cholesterol, steroids and lipids. CYP4F3, a 60 kDa protein, is a novel member of this family. It consists of a conserved heme-binding domain near the C-terminus and catalyzes the u-hydroxylation of LTB4. CYP4F3 is localized in the ER and plays a vital role in the inactivation of LTB4 and controlling inflammation. CYP4F3 exists in two functionally distinct isoforms: CYP4F3A, a neutrophil isoform, is particularly expressed in neutrophils and myeloid cells and catalyzes the microsomal u-hydroxylation of LTB4. It interacts with CD11b and myeloperoxidase on myeloid cells and regulates cellular development in bone marrow. CYP4F3B, a liver isoform, catalyzes the conversion of arachidonic acid to 20-HETE, a potent activator of PKC. Its role has been implicated in cell proliferation, vascular tone and natriuresis.Spezifikation
CYP4F3 | |
Polyclonal | |
Unconjugated | |
CYP4F3 | |
CPF3, CYP4F, CYP4F3, CYPIVF3, Cytochrome P450 4F3, Cytochrome P450 LTB omega, LTB4H | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4051 | |
-20°C | |
Liquid |
Western Blot | |
0.46 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q08477 | |
CYP4F3 | |
Peptide | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |