missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP4F11 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12286-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The cytochrome P450 proteins (CYPs) are monooxygenases that catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. P450 enzymes are classified into subfamilies based on their sequence similarities. CYP4F11 is an isoform of the cytochrome P450 4F protein and is expressed mainly in human liver, followed by kidney, heart, and skeletal muscle. CYP4F11 contains 524 amino acid residues that share 80.0, 82.3 and 79.2% identity to CYP4F2, CYP4F3, and CYP4F8 amino acid sequences, respectively. CYP4F11 has catalytic properties towards endogenous eicosanoids as well as some clinically relevant drugs, and is able to metabolize large molecules such as erythromycin.Spezifikation
CYP4F11 | |
Polyclonal | |
Unconjugated | |
CYP4F11 | |
CYP4F11, CYPIVF11, Cytochrome P450 4F11 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57834 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9HBI6 | |
CYP4F11 | |
CYP4F11 Fusion Protein Ag2906 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |