Learn More
Cyp2c7 Rabbit anti-Mouse, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 55452-1-AP-150UL
Beschreibung
The Cyp2c7 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of rat Cyp2c7. This antibody recognizes mouse, rat antigen. The Cyp2c7 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spezifikation
Cyp2c7 | |
Polyclonal | |
Unconjugated | |
150 μL | |
29298 | |
IgG |
Western Blot, Immunohistochemistry, ELISA | |
380 μg/mL | |
Rabbit | |
Primary | |
Mouse, Rat |