missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP1B1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 18505-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
CYP1B1 is a 543 amino acid intracellular protein localized to the endoplasmic reticulum. It belongs to the cytochrome P450 super family of enzymes which are monooxygenases and it catalyzes many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This enzyme metabolizes procarcinogens such as polycyclic aromatic hydrocarbons and 17beta-estradiol and is also involved in the metabolism of substances active in the eye growth and differentiation and hence plays a putative role in ocular development. Mutated proteins are known to cause primary congenital glaucoma.Spezifikation
CYP1B1 | |
Polyclonal | |
Unconjugated | |
CYP1B1 | |
CP1B, CYP1B1, CYPIB1, Cytochrome P450 1B1, GLC3A, P4501B1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13078, 1545, 25426 | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q16678, Q64429, Q64678 | |
Cyp1b1 | |
CYP1B1 Fusion Protein Ag13357 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Liquid |