missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Cynomolgus / Rhesus macaque TIGIT Protein, His Tag

Marke:  ACROBiosystems TIT-C5223-50UG

Additional Details : Gewicht : 0.00001kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15817777

  • 437.56 CHF / 50 Mikrogramm


Verfügbar ab
04-06-2021
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

T-cell immunoreceptor with Ig and ITIM domains (TIGIT) is also known as V-set and immunoglobulin domain-containing protein 9 (VSIG9), V-set and transmembrane domain-containing protein 3 (VSTM3),which belongs to single-pass type I membrane protein containing an immunoglobulin variable domain, a transmembrane domain and an immunoreceptor tyrosine-based inhibitory motif (ITIM). TIGIT is expressed at low levels on peripheral memory and regulatory CD4+ T-cells and NK cells and is up-regulated following activation of these cells (at protein level). TIGIT binds with high affinity to the poliovirus receptor (PVR) which causes increased secretion of IL10 and decreased secretion of IL12B and suppresses T-cell activation by promoting the generation of mature immunoregulatory dendritic cells.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.2 kDa. The protein migrates as 18-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation

Spezifikation

XP_015300911.1
HEK293
Cynomolgus / Rhesus macaque
15.2 kDa
50 µg
Met 89 - Ile 208
TIGIT,VSIG9,VSTM3
Unconjugated
PBS, pH7.4
Less than 1.0 EU / µg by the LAL method.
Immune Checkpoint Proteins
Powder
>90% as determined by SDS-PAGE
Cynomolgus / Rhesus macaque TIGIT, His Tag (TIT-C5223) is expressed from human 293 cells (HEK293). It contains AA Met 89 - Ile 208 (Accession # XP_015300911.1). In the region Met 89 - Ile 208, the AA sequence of Cynomolgus and Rhesus macaque TIGIT are homologus.
-20°C
TIGIT
His Tag
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation