Learn More
ACROBiosystems Cynomolgus / Rhesus macaque IL-4 R alpha / CD124 Protein, His Tag
466.58 CHF - 2912.92 CHF
Spezifikation
Molekulargewicht | 25.6 kDa |
---|---|
Quelle | Cynomolgus / Rhesus macaque IL-4 R alpha, His Tag (ILR-C52H8) is expressed from human 293 cells (HEK293). It contains AA Met 26 - Arg 232 (Accession # G7Q0S7). In the region Met 26 - Arg 232, the AA sequence of Cynomolgus and Rhesus macaque IL-4 R alpha are homologus. |
Immunogen | Met 26 - Arg 232 |
Lagerungsbedingungen | -20°C |
Gen-Alias | IL4R,CD124,IL4RA |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15827117
|
ACROBiosystems
ILR-C52H8-1MG |
1 mg |
2912.92 CHF
1 Milligramm |
Verfügbar ab: 16-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15837117
|
ACROBiosystems
ILR-C52H8-50UG |
50 μg |
466.58 CHF
50 Mikrogramm |
Verfügbar ab: 16-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
IL-4 is a pleiotropic cytokine produced by activated Th2 cells and mast cells, and plays a pivotal role in immune responses. The effects of IL-4 are mediated after binding to high affinity receptor complexes present on hematopoietic as well as non-hematopoietic cells. Hematopoietic cellular responses to IL-4 are mediated by a high affinity receptor complex comprised of the 140 kDa IL4Rα (CD124)subunit and the 70 kDa common cytokine γc chain (CD132).Interleukin 4 Receptor (IL4R) also known as CD124, IL4Rα and BSF receptor, is a type I cytokine receptor produced by activated Th2 cells and mast cells, and plays an important role in Th2- biased immune responses, alternative macrophage activation, mucosal immunity, allergic inflammation, tumor progression, and atherogenesis. A soluble form of the encoded IL4R protein can be produced by an alternate splice variant or by proteolysis of the membrane-bound protein, and this soluble form can inhibit IL4-mediated cell proliferation and IL5 upregulation by T-cells. IL4R can alternatively associate with IL-13Ra1 to form the type II receptor which is responsive to both IL-4 and IL-13. Interleukin-4 receptor has been shown to interact with SHC1.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.6 kDa. The protein migrates as 40-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
25.6 kDa | |
Met 26 - Arg 232 | |
IL4R,CD124,IL4RA | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
Cynomolgus / Rhesus macaque IL-4 R alpha, His Tag (ILR-C52H8) is expressed from human 293 cells (HEK293). It contains AA Met 26 - Arg 232 (Accession # G7Q0S7). In the region Met 26 - Arg 232, the AA sequence of Cynomolgus and Rhesus macaque IL-4 R alpha are homologus. | |
-20°C | |
IL-4 R alpha | |
Cynomolgus Monkey, Rhesus Macaque | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |