Learn More
ACROBiosystems Cynomolgus / Rhesus macaque B7-1 / CD80 Protein, His Tag
HPLC-verified
327.86 CHF - 2471.57 CHF
Spezifikation
Molekulargewicht | 25.8 kDa |
---|---|
Quelle | Cynomolgus / Rhesus macaque B7-1, His Tag (HPLC-verified) (CD0-C52H3) is expressed from human 293 cells (HEK293). It contains AA Val 35 - Asn 242 (Accession # G7NXN7). In the region Val 35 - Asn 242, the AA sequence of Cynomolgus and Rhesus macaque B7-1 are homologus. |
Immunogen | Val 35 - Asn 242 |
Lagerungsbedingungen | -20°C |
Gen-Alias | CD80,B7,B7-1,B7.1,BB1,CD28LG,CD28LG1,LAB7 |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15836117
|
ACROBiosystems
CD0-C52H3-1MG |
1 mg |
2471.57 CHF
1 Milligramm |
Verfügbar ab: 16-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15826117
|
ACROBiosystems
CD0-C52H3-100UG |
100 μg |
327.86 CHF
100 Mikrogramm |
Verfügbar ab: 16-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
B7-1 and B7-2, together with their receptors CD28 and CTLA-4, constitute one of the dominant co-stimulatory pathways that regulate T- and B-cell responses. Although both CTLA-4 and CD28 can bind to the same ligands, CTLA-4 binds to B7-1 and B7-2 with a 20 - 100 fold higher affinity than CD28 and is involved in the down regulation of the immune response.B-lymphocyte activation antigen B7-1 (referred to as B7) also known as cluster of Differentiation 80 (CD80), is a member of cell surface immunoglobulin superfamily and is expressed on activated B cells, activated T cells, macrophages and dendritic cells. It is the ligand for two different proteins on the T cell surface: CD28 (for autoregulation and intercellular association) and CTLA-4 (for attenuation of regulation and cellular disassociation). CD80 works in tandem with CD86 to prime T cells. CD80 plays a role in induction of innate immune responses by activating NF-κB-signaling pathway in macrophages. CD80 is thus regarded as promising therapeutic targets for autoimmune diseases and various carcinomas.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.8 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
25.8 kDa | |
Val 35 - Asn 242 | |
CD80,B7,B7-1,B7.1,BB1,CD28LG,CD28LG1,LAB7 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
Cynomolgus / Rhesus macaque B7-1, His Tag (HPLC-verified) (CD0-C52H3) is expressed from human 293 cells (HEK293). It contains AA Val 35 - Asn 242 (Accession # G7NXN7). In the region Val 35 - Asn 242, the AA sequence of Cynomolgus and Rhesus macaque B7-1 are homologus. | |
-20°C | |
B7-1 | |
Cynomolgus Monkey, Rhesus Macaque | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |