Learn More
ACROBiosystems Cynomolgus DR3 / TNFRSF25 Protein, Fc Tag
Marke: ACROBiosystems TN5-C5257-1MG
Additional Details : Gewicht : 0.00001kg
Beschreibung
Tumor necrosis factor receptor superfamily member 25 (TNFRSF25) is also known as Apo-3, Death receptor 3 (DDR3 or DR3), Apoptosis-inducing receptor AIR, Apoptosis-mediating receptor TRAMP, Lymphocyte-associated receptor of death, Apo-3, which is a member of the TNF-receptor superfamily. TNFRSF25 is a homodimer protein, which can Interact strongly via the death domains with TNFRSF1 and TRADD to activate at least two distinct signaling cascades, apoptosis and NF-kappa-B signaling. TNFRSF25 is receptor for TNFSF12 / APO3L / TWEAK.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 46.3 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
46.3 kDa | |
Cynomolgus DR3, Fc Tag (TN5-C5257) is expressed from human 293 cells (HEK293). It contains AA Gln 20 - Gly 203 (Accession # G7NTB5-1). | |
-20°C | |
DR3 / TNFRSF25 | |
Cynomolgus Monkey | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Other Recombinant Proteins |
1 mg | |
Gln 20 - Gly 203 | |
TNFRSF25,DR3,APO3,DDR3,TNFRSF12,WSL,WSL1 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |