Learn More
R&D Systems™ Cyno Monkey Integrin beta 5 (XP_005547990) VersaClone cDNA
Spezifikation
Spezifikation
| Zugriffsnummer | XP_005547990 |
| Gen-ID (Entrez) | 3693 |
| Inhalt und Lagerung | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date |
| Spezies | Cynomolgus Monkey |
| Menge | 10 μg |