Learn More
R&D Systems™ Cyno Monkey Glypican 3 (XP_005594665.1) VersaClone cDNA
Spezifikation
Spezifikation
| Gen | Glypican 3 |
| Zugriffsnummer | XP_005594665.1 |
| Konzentration | 10 μg at 0.2 μg/μL |
| Zusammensetzung | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Einsatztyp | 1756 bp |
| Gen-ID (Entrez) | 2719 |
| Gen-Alias | DGSX, glypican 3, glypican proteoglycan 3, glypican-3, GPC3, GTR2-2, heparan sulphate proteoglycan, Intestinal protein OCI-5, MXR7, OCI5, OCI-5, secreted glypican-3, SGB, SGBS, SGBS1SDYS |
| Inhalt und Lagerung | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Spezies | Cynomolgus Monkey |
| Menge | 10 μg |