missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin B1 Antibody (CCNB1/9242R), DyLight 650, Novus Biologicals™
Rabbit Monoclonal Antibody
Marke: Novus Biologicals NBP3-24309C
Dieser Artikel kann nicht zurückgegeben werden.
View return policy
Description
Cyclin B1 Monoclonal antibody specifically detects Cyclin B1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Cyclin B1 | |
| Monoclonal | |
| DyLight 650 | |
| CCNB, cyclin B1, G2/mitotic-specific cyclin B1, G2/mitotic-specific cyclin-B1 | |
| His-tagged recombinant hamster Cyclin B1 protein | |
| 0.1 mL | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cell Biology, Cell Cycle and Replication, Cytokine Research, Immunology, Mesenchymal Stem Cell Markers, Mitotic Regulators, Myeloid derived Suppressor Cell, Phospho Specific, Signal Transduction, Stem Cell Lines, Stem Cell Markers, Tumor Suppressors | |
| 891 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| CCNB1/9242R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction