missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CXCR4 Antibody (Dana-Farber patent anti-CXCR4), Novus Biologicals™
Human Monoclonal Antibody
248.00 CHF - 1080.00 CHF
Especificaciones
| Antigen | CXCR4 |
|---|---|
| Klon | Dana-Farber patent anti-CXCR4 |
| Anwendungen | Flow Cytometry, ELISA, Functional Assay |
| Klassifikation | Monoclonal |
| Konjugat | Unconjugated |
| Código de producto | Marca | Menge | Precio | Cantidad y disponibilidad | |||||
|---|---|---|---|---|---|---|---|---|---|
| Código de producto | Marca | Menge | Precio | Cantidad y disponibilidad | |||||
|
30047568
|
Novus Biologicals™
NBP3-28800-100ug |
100 μg |
394.00 CHF
100 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
30045105
|
Novus Biologicals™
NBP3-28800-1mg |
1 mg |
1080.00 CHF
1 Milligramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
30047060
|
Novus Biologicals™
NBP3-28800-50ug |
50 μg |
248.00 CHF
50 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Descripción
CXCR4 Monoclonal antibody specifically detects CXCR4 in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssayEspecificaciones
| CXCR4 | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Adaptive Immunity, Cancer, Chemokines and Cytokines, Cytokine Research, GPCR, Growth and Development, Immunology, Innate Immunity, Metastasis, Neuronal Cell Markers, Signal Transduction, Stem Cells | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CD184, chemokine (C-X-C motif) receptor 4, chemokine (C-X-C motif), receptor 4 (fusin), C-X-C chemokine receptor type 4, CXC-R4, CXCR-4, D2S201ESDF-1 receptor, FB22, Fusin, HM89NPYRL, HSY3RR, LAP3, LCR1, LESTRCD184 antigen, Leukocyte-derived seven transmembrane domain receptor, leukocyte-derived seven-transmembrane-domain receptor, lipopolysaccharide-associated protein 3, neuropeptide Y receptor Y3, NPY3RWHIM, NPYR, NPYY3R, pCXCR4, seven transmembrane helix receptor, seven-transmembrane-segment receptor, spleen, Stromal cell-derived factor 1 receptor | |
| CXCR4/ CD184 | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| Dana-Farber patent anti-CXCR4 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P61073 | |
| 7852 | |
| IgG1 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto