missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ CXCL1 (Human) Recombinant Protein (P01)

Human CXCL1 full-length ORF with GST-tag at N-terminal

Marke:  Abnova™ H00002919-P01.10ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16125791

  • 379.00 CHF / 10 Mikrogramm
Verfügbar ab: 09-01-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Human CXCL1 full-length ORF ( AAH11976, 1 a.a. - 107 a.a.) recombinant protein with GST-tag at N-terminal

  • Theoretical MW: 37.51kDa
  • Preparation Method: In vitro wheat germ expression system
  • Purification: Glutathione Sepharose 4 Fast Flow
  • Storage Buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer

Best use within three months from the date of receipt of this protein

Enzyme Linked Immunosorbent Assay, Western Blotting, Antibody Production, Protein Array

Spezifikation

Spezifikation

AAH11976
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
37.51
8
Glutathione Sepharose 4 Fast Flow
10 ug
MARAALSAAPSNPRLLRVALLLLLLVAAGRRAAGASVATELRCQCLQTLQGIHPKNIQSVNVKSPGPHCAQTEVIATLKNGRKACLNPASPIVKKIIEKMLNSDKSN
FSP/GRO1/GROa/MGSA/MGSA-a/NAP-3/SCYB1
CXCL1
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
2919
CXCL1 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CXCL1
Human
Recombinant
Solution
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote