Learn More
3M™ Culture Bottle RBTSB09036
Culture bottle for microbiological testing
Marke: 3M™ 7000053833
Additional Details : Gewicht : 0.34500kg
Beschreibung
- Culture Bottle
- Includes 90 mL Tryptic Soy Broth
- Recommended use For microbiological testing
Spezifikation
Food and Beverage, Environmental Test | |
90 mL Tryptic Soy Broth | |
36/Cs. |