Learn More
CTR2 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke:  Novus Biologicals NBP1-05199R
Artikelnummer. 18207260
  • 669.00 CHF / 0.10 Milliliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
CTR2 Polyclonal specifically detects CTR2 in Human, Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
CTR2
Polyclonal
Western Blot
O15432
SLC31A2
Synthetic peptide made to an internal portion of human CTR2 (within residues 50-100). [Swiss-Prot# O15432]
Affinity Purified
RUO
Primary
Human, Mouse
IgG
Western Blot
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
Copper transporter 2, COPT2SLC13A2, CTR2Solute carrier family 31 member 2, hCTR2probable low affinity copper uptake protein 2, solute carrier family 31 (copper transporters), member 2
Rabbit
71 kDa
0.1 mL
Cancer, Lipid and Metabolism, Plasma Membrane Markers, Signal Transduction
1318
Store at 4C in the dark.
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen