missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CTBP2 (Human) Recombinant Protein
Human CTBP2 full-length ORF ( AAH02486, 1 a.a. - 445 a.a.) recombinant protein with GST-tag at N-terminal.
Marke: Abnova™ H00001488-P01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
- Sequence: MALVDKHKVKRQRLDRICEGIRPQIMNGPLHPRPLVALLDGRDCTVEMPILKDLATVAFCDAQSTQEIHEKVLNEAVGAMMYHTITLTREDLEKFKALRVIVRIGSGYDNVDIKAAGELGIAVCNIPSAAVEETADSTICHILNLYRRNTWLYQALREGTRVQSVEQIREVASGAARIRGETLGLIGFGRTGQAVAVRAKAFGFSVIFYDPYLQDGIERSLGVQRVYTLQDLLYQSDCVSLHCNLNEHNHHLINDFTIKQMRQGAFLVNAARGGLVDEKALAQALKEGRIRGAALDVHESEPFSFAQGPLKDAPNLICTPHTAWYSEQASLEMREAAATEIRRAITGRIPESLRNCVNKEFFVTSAPWSVIDQQAIHPELNGATYRYPPGIVGVAPGGLPAAMEGIIPGGIPVTHNLPTVAHPSQAPSPNQPTKHGDNREHPNEQ
Spezifikation
AAH02486 | |
C-terminal binding protein 2 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
1488 | |
Wheat germ expression system | |
25 μg | |
CTBP2 | |
GST |