Learn More
Invitrogen™ CSNK1G3 (CK1 γ 3) Recombinant Human Protein
The casein kinase I (CKI) gene family encodes serine/threonine kinases that preferentially phosphorylate acidic substrates.
Marke: Invitrogen™ PV3991
Additional Details : Gewicht : 0.00900kg
Beschreibung
CKI proteins are monomeric and range from 25 to 55 kD. They are ubiquitous, being found in the nuclei, cytoplasm, and membrane fractions of eukaryotic cells
Spezifikation
NP_004375, R174G | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
1mg | |
RUO - research use only | |
CSNK1G3 (CK1 γ 3) | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
78.9kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
CK1 Kinase Family | |
Recombinant | |
Baculovirus |