Learn More
CSN1S1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 22648-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The CSN1S1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human CSN1S1. This antibody recognizes human antigen. The CSN1S1 antibody has been validated for the following applications: WB, ELISA analysis.
Spezifikation
| CSN1S1 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 1446 | |
| IgG |
| Western Blot, ELISA | |
| 400 μg/mL | |
| Rabbit | |
| Primary | |
| Human |