missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cryptosporidium Antibody (BEL 0126), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NB100-64526PECY55
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Cryptosporidium Monoclonal specifically detects Cryptosporidium in Protozoa samples. It is validated for Flow Cytometry.
Spezifikation
| Cryptosporidium | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| Mouse | |
| Protein A purified | |
| Primary | |
| Protozoa | |
| IgG3 |
| Flow Cytometry | |
| BEL 0126 | |
| Flow Cytometry | |
| Cryptosporidiosis | |
| Purified oocysts from bovine faeces | |
| 0.1 mL | |
| Recognizes a membrane antigen expressed by the oocysts of Cryptosporidium sp., an obligate intracellular parasite of the phylum Apicomplexa. The genome of Cryptosporidium sp. is one of the smallest among unicellular eukaryotic organisms. Cryptosporidium is one of several species that cause cryptosporidiosis; a disease affecting the intestines of mammals which is spread through the faecal-oral route. The main symptom is self-limiting diarrhoea in people with intact immune systems. However, in immunocompromised individuals, such as AIDS patients, infection can cause permanent & life-threatening diarrhoea. Cryptosporidiosis is one of the most common waterborne diseases worldwide. | |
| Store at 4°C in the dark. Do not freeze. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur