missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRTAC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 13001-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
CRTAC1 encodes a glycosylated extracellular matrix protein that is found in the interterritorial matrix of articular deep zone cartilage. This protein is used as a marker to distinguish chondrocytes from osteoblasts and mesenchymal stem cells in culture. The presence of FG-GAP motifs and an RGD integrin-binding motif suggests that this protein may be involved in cell-cell or cell-matrix interactions. Copy number alterations in this gene have been observed in neurofibromatosis type 1-associated glomus tumors. Alternative splicing results in multiple transcript variants.Spezifikation
CRTAC1 | |
Polyclonal | |
Unconjugated | |
CRTAC1 | |
ASPIC, ASPIC1, cartilage acidic protein 1, CEP 68, CEP68, CRTAC1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
171438, 55118, 72832 | |
-20°C | |
Liquid |
Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8R555, Q9NQ79 | |
Crtac1 | |
CRTAC1 Fusion Protein Ag3673 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |