Beschreibung
- With cross beater, baffle plates, 240 mm filter hose and collecting receptacle
- 3-phase motor with frequency converter
- Suitable for batchwise operation of larger quantities
- Adjustable speed for different applications
- Defined final fineness due to bottom sieves with aperture sizes from 0.12 - 10 mm
- Easy cleaning thanks to exchangeable push-fit grinding insert, rotor and sieve
- Wide range of accessories including various collection and feeding systems, rotors and sieves
- Electric engine brake
- Central locking device
- Optional cyclone available
Spezifikation
Spezifikation
| Abmessungen (L x B x H) | 700 x 600 x 1200 mm |
| Elektrische Anforderungen | 200/240 V, 50/60 Hz |
| Enthält | Prallplatten aus Stahl 1.1740 |
| Material | Gusseisen |
| Phase | 1-phase connection |
| Geschwindigkeit | 2000 to 4000 rpm |