missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRISPR-Cas9 Antibody Pack, Novus Biologicals™
Mouse Antibody
Marke: Novus Biologicals NBP2-52986
Additional Details : Gewicht : 0.58200kg
Beschreibung
CRISPR-Cas9 specifically detects CRISPR-Cas9 in Bacteria samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Spezifikation
CRISPR-Cas9 | |
Unconjugated | |
Cas9, CRISPR, CRISPR-associated protein 9 nuclease, SpyCas9 | |
NBP2-36440 = Recombinant N-terminal fragment of S.pyogenes CRISPR/Cas9. [UniProt# Q99ZW2]NBP2-52398 = Recombinant C-terminal fragment of S.pyogenes CRISPR/Cas9. [UniProt# Q99ZW2] | |
Gene Editing, Gene Knockouts, Gene Therapy | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP) | |
Mouse | |
2 Vials | |
Bacteria |