Learn More
CRHR1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20967-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This antibody raised against a synthesized peptide corresponding to amino acid residues 148-165 of isoform CRF-R1 can only recognize isoform CRF-R1.
Corticotropin-releasing factor receptor 1 (CRFR1/CRHR1), a Corticotropin-Releasing Factor Receptor, is involved in mediating the stress response and anxiety-related behaviors. Mutations of the receptor reduce anxiety-related behaviors and may be involved in the pathogenesis of Cushing disease. Multiple isoforms of CRFR1 are produced by alternative splicing. CRFR1 has been reported to be expressed in the pituitary, adrenal gland, brain, colon, ovary, placenta, testis, white blood cells, and skin. ESTs have been isolated from human brain, eye, colon, and embryo libraries.Spezifikation
CRHR1 | |
Polyclonal | |
Unconjugated | |
CRHR1 | |
CRF R, CRF R 1, CRF R1, CRF1, CRFR, CRFR 1, CRFR1, CRH R 1, CRH R1, CRH R1h, CRHR, CRHR1, CRHR1f | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1394 | |
Antibody | |
IgG |
Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P34998 | |
CRHR1 | |
Peptide | |
150 μL | |
Primary | |
Human | |
Liquid |