Learn More
Creatine Kinase, Muscle/CKMM Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
418.00 CHF - 668.00 CHF
Spezifikation
| Antigen | Creatine Kinase, Muscle/CKMM |
|---|---|
| Verdünnung | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Anwendungen | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18439301
|
Novus Biologicals
NBP2-13839 |
0.1 mL |
668.00 CHF
0.10 Milliliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18421492
|
Novus Biologicals
NBP2-13839-25ul |
25 μL |
418.00 CHF
25 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Creatine Kinase, Muscle/CKMM Polyclonal antibody specifically detects Creatine Kinase, Muscle/CKMM in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Spezifikation
| Creatine Kinase, Muscle/CKMM | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| CKMMcreatine kinase M-type, Creatine Kinase Isoenzyme, Creatine kinase M chain, creatine kinase, muscle, creatine kinase-M, EC 2.7.3, EC 2.7.3.2, M-CK | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: MPFGNTHNKFKLNYKPEEEYPDLS | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol | |
| 1158 | |
| IgG | |
| Immunogen affinity purified |