missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CPT 11
GREENER_CHOICE

Artikelnummer. p-4783884
Click to view available options
Menge:
10 mg
50 mg
missing translation for 'unitSize'
10 Milligramm
50 Milligramm
This item is not returnable. View return policy

Product Code. 18752534

missing translation for 'mfr': Tocris Bioscience™ 2688/10

om dit product te kopen Registreer vandaag om een webaccount aan te maken

This item is not returnable. View return policy

DNA topoisomerase I inhibitor; antitumor

Inhibitor of DNA topoisomerase I that displays antitumor activity against a range of tumor types. Active metabolite available ( SN 38, Cat. No. 2684).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100286-90-6
Summenformel C33H39ClN4O6

Specifications

Chemischer Name oder Material CPT 11
Menge 10 mg
Formelmasse 623.14
Reinheit >99%
Molekulargewicht 623.15
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.