missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX6C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11429-2-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Cytochrome c oxidase, the terminal enzyme of the mitochondrial respiratory chain, catalyzes the electron transfer from reduced cytochrome c to oxygen. It is a heteromeric complex consisting of 3 catalytic subunits encoded by mitochondrial genes and multiple structural subunits encoded by nuclear genes. The mitochondrially-encoded subunits function in electron transfer, and the nuclear-encoded subunits may be involved in the regulation and assembly of the complex. This nuclear gene encodes subunit VIc, which has 77% amino acid sequence identity with mouse subunit VIc. This gene is up-regulated in prostate cancer cells. A pseudogene has been found on chromosomes 16p12.Spezifikation
COX6C | |
Polyclonal | |
Unconjugated | |
COX6C | |
COX6C | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12864, 1345, 54322 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P09669, P11951, Q9CPQ1 | |
Cox6c | |
COX6C Fusion Protein Ag1982 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |