missing translation for 'onlineSavingsMsg'
Learn More

COX5B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 11418-2-AP-150UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16838483

  • 351.68 CHF / 150 Mikroliter

Verfügbar ab: 28-06-2022
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The cytochrome c oxidase (COX) family of proteins function as the final electron donor in the respiratory chain to drive a proton gradient across the inner mitochondrial membrane, ultimately resulting in the production of water. The mammalian COX apoenzyme is a dimer, with each monomer consisting of 13 subunits, some of which are mitochondrial and some of which are nuclear. Found in the inner mitochondrial membrane, COX5 is the heme A-containing chain of the oxidase family that converts one molecule of oxygen and four molecules of hydrogen to two molecules of water. Two isoforms of COX5 exist, COX5a and COX5b. When oxygen levels within the cell are high, transcription of COX5a (the aerobic isoform) is up-regulated as the rate of cellular respiration increases. Conversely, when oxygen levels are low, COX5b (the hypoxic isoform) transcription increases and functions to maximize the turnover rate of the COX apoenzyme.
Spezifikation

Spezifikation

COX5B
Polyclonal
Unconjugated
COX5B
COX5B, COXVB
Rabbit
Antigen Affinity Chromatography
RUO
12859, 1329, 94194
Antibody
IgG
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P10606, P12075, P19536
Cox5b
COX5B Fusion Protein Ag1986
150 μL
Primary
Human, Mouse, Rat
Liquid
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate
Sonderangebote

Sonderangebote