Learn More
SNV Comines™ Cotton Laboratory Coats
Product Code. 12120920
Change view
Click to view available options
Size:
42
44
50
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12120920 50 Each
12150920 42 Each
12160920 44 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12120920 Supplier SNV Comines™ Supplier No. ANTLB00400T3
  • ONLINE EXCLUSIVE PRICE
    33.55 CHF
    18.26 CHF / 1 Stück
    Sparen 15.29 CHF
    45% Rabatt
Lager vorrätig
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Provides general light protection against splashes in the laboratory. SNV Comines™ Cotton Laboratory Coats are available in a range of sizes.
  • White in color
  • Collared with lapels
  • 4 buttons
Specifications
Product Type Xavier Lab Coat
Size 50
Gender Men's
Material Carbon
Color White
Closure Type Buttons
No. of Pockets 4
Size (Metric) Chest 108 cm
Includes 2 Flat pockets, 1 breast pocket, 1 inside pocket, opening angle
Construction 100% Cotton