Learn More
CoraLite™ Plus 488-conjugated CD8a Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech CL488-66868-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The CD8a antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD8a. This antibody recognizes human antigen. The CD8a antibody has been validated for the following applications: IF-P analysis. This antibody is coralite™ plus 488.Spezifikation
| CD8a | |
| 1G2B10 | |
| CoraLite Plus 488 | |
| Mouse | |
| Protein G purification | |
| Primary | |
| Human | |
| Conjugated Antibody | |
| IgG1 |
| Immunofluorescence | |
| 1000 μg/mL | |
| CD8; Leu2; MAL; p32 | |
| Recombinant Protein | |
| 20 μL | |
| 925 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |