missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kupfer(II)-sulfat-Pentahydrat, Puriss., geeignet für, gemäß analytischen Spezifikationen von BP, FCC, Europ. Arzneibuch, USP, Honeywell Fluka™
Click to view available options
Menge:
100 g
250 g
500 g
1 kg
2.5 kg
6 x 500 g
25 kg
6 x 1 kg
6 x 2.5 kg
Beschreibung
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Spezifikation
Chemischer Name oder Material | Copper(Ii) Sulfate Pentahydrate |
Namenshinweis | Meets analytical specification of Ph. Eur.BP, USP, FCC |
Menge | 6 x 500 g |
Strukturformel | CuSO4 · 5H2O |
UN-Nummer | 3077 |
IUPAC-Name | copper(2+) pentahydrate sulfate |
Formelmasse | 249.69g/mol |
Güte | puriss. |
Dampfdruck | 7.3mmHg (25°C) |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'