missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COPG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | COPG |
---|---|
Konzentration | 0.13 mg/mL |
Anwendungen | Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16850094
|
Proteintech
12393-1-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16840094
|
Proteintech
12393-1-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The coatomer is a cytosolic protein complex that binds to dilysine motifs and reversibly associates with Golgi non-clathrin-coated vesicles, which further mediate biosynthetic protein transport from the ER, via the Golgi up to the trans Golgi network. Coatomer complex is required for budding from Golgi membranes, and is essential for the retrograde Golgi-to-ER transport of dilysine-tagged proteins. In mammals, the coatomer can only be recruited by membranes associated to ADP-ribosylation factors (ARFs), which are small GTP-binding proteins; the complex also influences the Golgi structural integrity, as well as the processing, activity, and endocytic recycling of LDL receptors.Spezifikation
COPG | |
Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
Q4AEF8, Q9QZE5, Q9Y678 | |
22820, 297428, 54161 | |
COPG Fusion Protein Ag3070 | |
Primary | |
-20°C | |
COPG1 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Coatomer subunit gamma, COPG, COPG1, Gamma coat protein, Gamma COP | |
Copg1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |