Learn More
WTW™ ConOx-1.5 Leitfähigkeits-/Sauerstoffsensor 1,5m Kabel, komplett mit Zubehör
Artikelnummer. 10627113
Change view
Click to view available options
Länge (metrisch) Kabel:
1,5 m
Packungsgröße:
1 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Länge (metrisch) Kabel unitSize
10627113 1,5 m 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 10627113 Lieferant WTW™ Lieferanten-Nr. 401010
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen
Spezifikation
Typ Combined Oxygen/Conductivity Sensor
Länge (metrisch) Kabel 1,5 m
Material Epoxidharz/Graphit
Enthält 1.5m L Cable, with 3 exchange membrane heads, cleaning solution, conductivity standard (1413μS/cm), abrasive foil, OxiCal™ CX calibration vessel, electrolyte
Modell ConOx-1.5