Learn More
Thermo Scientific™ Collagenase, Type I, Clostridium histolyticum
Type I Collagenase (CAS # 9001-12-1) is one of seven collagenases, enzymes responsible for collagen degradation.
auf Anfrage
Beschreibung
Beschreibung
Collagenase, Type I, Clostridium histolyticum is used as a powder and solvent injection kit for the treatment of dupuytren's contracture.
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
General Description
  • Type I collagenase is a collagen-specific enzyme required for the proper degradation of connective tissue
  • Type I collagenase is isolated from Clostridium histolyticum
  • It is implicated in connective tissue disorders such as Peyronie’s and Dupuytren’s
Application
  • Type I collagenase targets specific alpha helices within collagen
  • It is suitable for use in cell culture and biochemistry applications
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Product Certifications
RUO – Research Use Only