Learn More
Novus Biologicals™ CLOCK Overexpression Lysate
Marke:  Novus Biologicals™ NBP2-07811
Additional Details : Gewicht : 0.00970kg
Artikelnummer. 18007677
  • 901.70 CHF / 0.10 Milligramm
Verfügbar ab: 17-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
The CLOCK Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Spezifikation
Spezifikation
Western Blot
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_4889. The protein contains a C-terminal DDK tag.
RIPA buffer
Homo sapiens CLOCK, mRNA.
Native