Learn More
Novus Biologicals™ CLOCK Overexpression Lysate
Marke: Novus Biologicals™ NBP2-07811
Additional Details : Gewicht : 0.00970kg
Beschreibung
The CLOCK Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.Spezifikation
Western Blot | |
0.1 mg | |
Protein | |
-80°C. Avoid freeze-thaw cycles. |
The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_4889. The protein contains a C-terminal DDK tag. | |
RIPA buffer | |
Homo sapiens CLOCK, mRNA. | |
Native |
For Research Use Only