missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Clobetasol Propionate Monoclonal Antibody (1B7)

Mouse Monoclonal Antibody
Marke: Invitrogen™ MA549727
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Antibody tested in indirect ELISA.
Clobetasol Propionate is a synthetic corticosteroid used to treat skin conditions such as eczema, psoriasis, and dermatitis. It works by reducing inflammation, itching, and redness in affected areas. It is available as a cream, ointment, or lotion and is applied topically to the affected area. Clobetasol Propionate should be used sparingly and only as directed by a healthcare professional, as prolonged use can lead to skin thinning and other adverse effects.
Spezifikation
| Clobetasol Propionate | |
| Monoclonal | |
| 1.75 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Conjugate of CBP hapten and KLH. | |
| 200 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry, Lateral Flow, SDS-Page | |
| 1B7 | |
| Unconjugated | |
| Mouse | |
| Protein G | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur