missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLN6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20315-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
CLN6, a 311-amino acid protein, has 7 predicted transmembrane domains and is conserved across vertebrates. The CLN6 protein localizes to the endoplasmic reticulum but contributes to lysosomal function. Mutations in the CLN6 gene cause variant late-onset infantile neuronal ceroid lipofuscinosis (vLINCL), a lysosomal storage disorder marked by progressive mental deterioration and blindness; part of a group of severe inherited neurodegenerative disorders affecting children wherein lysosomes accumulate storage material, causing the death of neurons. CLN6 is one of eight proteins, including CLN1-8, that are associated with NCL.Spezifikation
CLN6 | |
Polyclonal | |
Unconjugated | |
CLN6 | |
CLN6, HsT18960, nclf, Protein CLN6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54982 | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.51 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9NWW5 | |
CLN6 | |
CLN6 Fusion Protein Ag14159 | |
20 μL | |
Primary | |
Human | |
Liquid |