Learn More
CLK4 Recombinant Human Protein
CLK4 Recombinant Human Protein
Marke:  Invitrogen PV3839
Additional Details : Gewicht : 0.05250kg
Artikelnummer. 11513577
  • 850.00 CHF / 10 Mikrogramm
Lager vorrätig
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
CLK4 belongs to the CDC2-like protein kinase (CLK) family. It can interact with and phosphorylate the serine- and arginine-rich (SR) proteins, which are known to play an important role in the formation of spliceosomes, and thus is thought to be involved in the regulation of alternative splicing.
Spezifikation
Spezifikation
NP_065717
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay
85kDa
10μg
For research use only. Not for use in diagnostic procedures
CLK4
CLK4 was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 100μM RS domain derived peptide substrate (GRSRSRSRSR) and trace [32P]-γ-ATP for 10 minutes at 30°C
CLK4 Recombinant Human Protein
Human
Baculovirus
Serine/threonine kinase
0.24mg/mL
57396, 7040
CLK4 Recombinant Human Protein
–80°C
1 Each
CLK4 Recombinant Human Protein
CLK
44nmoles of phosphate transferred to RS domain derived peptide substrate (GRSRSRSRSR) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 8.33μg/mL
GST Tag
Kinase and inhibitors
90% by SDS-PAGE
Produktvorschläge
Produktvorschläge
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only. Not for use in diagnostic procedures.